Anti-ITPR3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20248
10719-918EA 428.55 USD
10719-918
Anti-ITPR3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant ITPR3.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500

Type: Primary
Antigen: ITPR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR