Anti-AHR Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12490
10688-556EA 653.85 USD
10688-556
Anti-AHR Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against partial recombinant AHR.

Recommended Dilutions: ELISA: 1:100; Immunocytochemistry: 1:100; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:100

Type: Primary
Antigen: AHR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Guinea Pig, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR