Anti-E2F7 Goat Polyclonal Antibody

Supplier: Abnova
PAB12815
10711-560EA 428 USD
10711-560
Anti-E2F7 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of E2F7.

Recommended Dilutions: ELISA: 1:16000; Western Blot: 0.1-0.3 ?g/ml

Type: Primary
Antigen: E2F7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR