Anti-TNFRSF10D Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12831
10742-064EA 428 USD
10742-064
Anti-TNFRSF10D Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of TNFRSF10D.

Recommended Dilutions: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: TNFRSF10D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR