Anti-IKBKB Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12836
10721-226EA 428 USD
10721-226
Anti-IKBKB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of IKBKB.

Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: IKBKB
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR