Anti-TMX4 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20845
10743-428EA 428.55 USD
10743-428
Anti-TMX4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant TXNDC13.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Western Blot: 1:250-1:500

Type: Primary
Antigen: TMX4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR