Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: DR5-01]

Supplier: Abnova
MAB2526
10741-450EA 638 USD
10741-450
Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: DR5-01]
Antibodies
Mouse monoclonal antibody raised against partial recombinant TNFRSF10B.

Type: Primary
Antigen: TNFRSF10B
Clonality: Monoclonal
Clone: DR5-01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR