Anti-TMEM45B Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20853
10741-884EA 428.55 USD
10741-884
Anti-TMEM45B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant TMEM45B.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: TMEM45B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR