Anti-IRAK4 Mouse Monoclonal Antibody [clone: A8A8]

Supplier: Abnova
MAB2538
10681-702EA 346 USD
10681-702
Anti-IRAK4 Mouse Monoclonal Antibody [clone: A8A8]
Antibodies
Mouse monoclonal antibody raised against synthetic phosphopeptide of IRAK4.

Recommended Dilutions: Western Blot: 0.1-1 ?g/ml; ELISA: 0.01-0.1 ?g/ml; Immunoprecipitation: 2-5 ?g/ml; Immunohistochemistry: 2-5 ?g/ml

Type: Primary
Antigen: IRAK4
Clonality: Monoclonal
Clone: A8A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Bovine, Dog, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR