Anti-CK18 Mouse Monoclonal Antibody [clone: SPM265]

Supplier: Abnova
MAB2540
10722-896EA 329 USD
10722-896
Anti-CK18 Mouse Monoclonal Antibody [clone: SPM265]
Antibodies
Mouse monoclonal antibody raised against native KRT18.

Recommended Dilutions: Western Blot: 0.1-1 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:50: 2-5 ?g/ml

Type: Primary
Antigen: CK18
Clonality: Monoclonal
Clone: SPM265
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR