Anti-IKBKAP Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12857
10718-946EA 428 USD
10718-946
Anti-IKBKAP Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of IKBKAP.

Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: IKBKAP
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR