Anti-COL5A1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13507
10708-746EA 531 USD
10708-746
Anti-COL5A1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against native COL5A1/COL5A2/COL5A3.

Recommended Dilutions: ELISA: 1:200; Immunohistochemistry: 1:500; Immunofluorescence: 1:80

Type: Primary
Antigen: COL5A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR