Anti-CASP7 Mouse Monoclonal Antibody [clone: MCH3-14]

Supplier: Abnova
MAB3467
10705-648EA 849 USD
10705-648
Anti-CASP7 Mouse Monoclonal Antibody [clone: MCH3-14]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of CASP7.

Type: Primary
Antigen: CASP7
Clonality: Monoclonal
Clone: MCH3-14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR