Anti-FN1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13518
10734-990EA 1170 USD
10734-990
Anti-FN1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against native FN1.

Recommended Dilutions: Immunohistochemistry: 1:200-1:600; Immunofluorescence: 1:40

Type: Primary
Antigen: FN1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR