Anti-CASP2 Rat Monoclonal Antibody [clone: 10C6]

Supplier: Abnova
MAB3484
10705-432EA 1169.39 USD
10705-432
Anti-CASP2 Rat Monoclonal Antibody [clone: 10C6]
Antibodies
Rat monoclonal antibody raised against recombinant CASP2.

Type: Primary
Antigen: CASP2
Clonality: Monoclonal
Clone: 10C6
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR