Anti-Hemoglobin Mouse Monoclonal Antibody [clone: 902]

Supplier: Abnova
MAB4185
10676-422EA 2295.84 USD
10676-422
Anti-Hemoglobin Mouse Monoclonal Antibody [clone: 902]
Antibodies
Mouse monoclonal antibody raised against native Hemoglobin.

Type: Primary
Antigen: Hemoglobin
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 902
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR