Anti-HOMER1 Mouse Monoclonal Antibody [clone: AT1F3]

Supplier: Abnova
MAB4202
10717-744EA 329 USD
10717-744
Anti-HOMER1 Mouse Monoclonal Antibody [clone: AT1F3]
Antibodies
Mouse monoclonal antibody raised against full length recombinant HOMER1.

Type: Primary
Antigen: HOMER1
Clonality: Monoclonal
Clone: AT1F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR