Anti-CGB Mouse Monoclonal Antibody [clone: 057-10020]

Supplier: Abnova
MAB4210
10626-978EA 530.32 USD
10626-978
Anti-CGB Mouse Monoclonal Antibody [clone: 057-10020]
Antibodies
Mouse monoclonal antibody raised against native CGB.

Type: Primary
Antigen: CGB
Clonality: Monoclonal
Clone: 057-10020
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR