Anti-ABCG8 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12502
10673-092EA 530.32 USD
10673-092
Anti-ABCG8 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of ABCG8.

Recommended Dilutions: Western Blot: 1:250

Type: Primary
Antigen: ABCG8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR