Anti-ABCA1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12504
10687-584EA 531 USD
10687-584
Anti-ABCA1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of ABCA1.

Recommended Dilutions: Flow Cytometry: 1:200; Immunocytochemistry: 1:200; Immunofluorescence: 1:200; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:200; Immunoprecipitation: 1:500; Western Blot: 1:500

Type: Primary
Antigen: ABCA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Hamster, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR