Anti-Influenza A virus Haemagglutinin H5 (Avian) Mouse Monoclonal Antibody [clone: B205M]

Supplier: Abnova
MAB4827
10716-880EA 956.86 USD
10716-880
Anti-Influenza A virus Haemagglutinin H5 (Avian) Mouse Monoclonal Antibody [clone: B205M]
Antibodies
Mouse monoclonal antibody raised against Influenza A Virus Haemagglutinin H5 (Avian).

Recommended Dilutions: ELISA: detect 10ng of free peptide at 1 ?g/ml

Type: Primary
Antigen: Influenza A virus Haemagglutinin H5 (Avian)
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: B205M
Epitope:
Host: IgG
Isotype: Mouse
Reactivity: Viruses
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR