Anti-BCL10 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12511
10690-904EA 638.29 USD
10690-904
Anti-BCL10 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against partial recombinant BCL10.

  • Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: BCL10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR