Anti-SF4 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20289
10738-160EA 428.55 USD
10738-160
Anti-SF4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant SF4.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Western Blot: 1:250-1:500; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: SF4
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR