Anti-CYR61 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12525
10712-578EA 530.32 USD
10712-578
Anti-CYR61 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of CYR61.

Recommended Dilutions: Western Blot: 1:200; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:100

Type: Primary
Antigen: CYR61
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR