Anti-NR1H3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12535
10733-126EA 530.32 USD
10733-126
Anti-NR1H3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of NR1H3/NR1H2.

Recommended Dilutions: Western Blot: 0.5-2 ?g/ml

Type: Primary
Antigen: NR1H3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Human, Mouse, Primates, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR