Anti-PERP Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12866
10728-974EA 428 USD
10728-974
Anti-PERP Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of PERP.

Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: PERP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR