Anti-BIRC7 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12872
10691-314EA 428 USD
10691-314
Anti-BIRC7 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of BIRC7.

Recommended Dilutions: Western Blot: 2-4 ?g/ml

Type: Primary
Antigen: BIRC7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR