Anti-NLRC4 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12895
10726-592EA 428 USD
10726-592
Anti-NLRC4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of NLRC4.

Recommended Dilutions: Western Blot: 0.5-2 ?g/ml

Type: Primary
Antigen: NLRC4
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR