Anti-BCL2L2 Rat Monoclonal Antibody [clone: 13F9]

Supplier: Abnova
MAB3487
10691-014EA 1061.25 USD
10691-014
Anti-BCL2L2 Rat Monoclonal Antibody [clone: 13F9]
Antibodies
Rat monoclonal antibody raised against full length recombinant BCL2L2.

Type: Primary
Antigen: BCL2L2
Clonality: Monoclonal
Clone: 13F9
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR