Anti-VTG Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13582
10677-464EA 638 USD
10677-464
Anti-VTG Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against full length native Vitellogenin (VTG).

Recommended Dilutions: Western Blot: 1:5000; ELISA: 1:5000

Type: Primary
Antigen: VTG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: European seabass, Salmonids, Senegalese sole
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR