Anti-RSAD1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21496
10732-050EA 527.69 USD
10732-050
Anti-RSAD1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant RSAD1.

Recommended Dilutions: Immunohistochemistry: 1:20-1:50

Type: Primary
Antigen: RSAD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR