Anti-MUG1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13585
10676-886EA 786.15 USD
10676-886
Anti-MUG1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against Mug1.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: MUG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR