Anti-APP Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13588
10691-130EA 530.63 USD
10691-130
Anti-APP Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Beta amyloid.

  • Recommended Dilutions: ELISA: 1:3000; Dot Blot: 1:1000; Western Blot: 1:1000

Type: Primary
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR