Anti-NEFM Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21508
10732-962EA 428.55 USD
10732-962
Anti-NEFM Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant NEFM.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000

Type: Primary
Antigen: NEFM
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR