Anti-13(S)-HODE Polyclonal Antibody

Supplier: Abnova
PAB13604
10687-362EA 427.45 USD
10687-362
Anti-13(S)-HODE Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against 13(S)-HODE.

Recommended Dilutions: Immunohistochemistry: 2 ?g/well; Enzyme Immunoassay: 2 ?g/well; Radio Immunoassay: 2 ?g/well)

Type: Primary
Antigen: 13(S)-HODE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR