Anti-ALOX8 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13607
10688-846EA 530.32 USD
10688-846
Anti-ALOX8 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against Alox8.

Recommended Dilutions: Western Blot: 1:3000

Type: Primary
Antigen: ALOX8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR