Anti-Epstein-Barr virus Mouse Monoclonal Antibody [clone: CS1-4 Cocktail]

Supplier: Abnova
MAB3534
10627-346EA 531 USD
10627-346
Anti-Epstein-Barr virus Mouse Monoclonal Antibody [clone: CS1-4 Cocktail]
Antibodies
Mouse monoclonal antibody raised against recombinant Epstein-Barr virus.

Recommended Dilutions: Immunohistochemistry: 1:10-1:20

Type: Primary
Antigen: Epstein-Barr virus
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: CS1-4 Cocktail
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR