Anti-RC3H1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB22034
10734-182EA 428.55 USD
10734-182
Anti-RC3H1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant RC3H1.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500

Type: Primary
Antigen: RC3H1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR