Anti-CGN Rabbit Polyclonal Antibody

Supplier: Abnova
PAB22059
10710-138EA 428.55 USD
10710-138
Anti-CGN Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant CGN.

Recommended Dilutions: Immunohistochemistry: 1:1000-1:2500; Western Blot: 1:250-1:500

Type: Primary
Antigen: CGN
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR