Anti-CD300E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: UP-H2]

Supplier: Abnova
MAB4998
10706-036EA 530.63 USD
10706-036
Anti-CD300E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: UP-H2]
Antibodies
Mouse monoclonal antibody raised against CD300E.

Recommended Dilutions: Flow Cytometry: 20 ?l/10^6 cells

Type: Primary
Antigen: CD300E
Clonality: Monoclonal
Clone: UP-H2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR