Anti-NCAM1 Mouse Monoclonal Antibody (Biotin) [clone: MEM-188]

Supplier: Abnova
MAB5002
10726-686EA 329 USD
10726-686
Anti-NCAM1 Mouse Monoclonal Antibody (Biotin) [clone: MEM-188]
Antibodies
Mouse monoclonal antibody raised against native NCAM1.

Recommended Dilutions: Flow Cytometry: 5 ?g/ml

Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: MEM-188
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Primates
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR