Anti-CD58 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-63]

Supplier: Abnova
MAB5007
10706-704EA 329 USD
10706-704
Anti-CD58 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-63]
Antibodies
Mouse monoclonal antibody raised against native CD58.

Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)

Type: Primary
Antigen: CD58
Clonality: Monoclonal
Clone: MEM-63
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR