Anti-GRHL1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20320
10716-398EA 428.55 USD
10716-398
Anti-GRHL1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant GRHL1.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Western Blot: 1:250-1:500; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: GRHL1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR