Anti-CD59 Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-43]

Supplier: Abnova
MAB5008
10706-712EA 329 USD
10706-712
Anti-CD59 Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-43]
Antibodies
Mouse monoclonal antibody raised against native CD59.

Recommended Dilutions: Flow Cytometry: 10 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)

Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: MEM-43
Conjugation: APC (Allophycocyanin)
Epitope:
Host:
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR