Anti-TFRC Mouse Monoclonal Antibody (Biotin) [clone: MEM-75]

Supplier: Abnova
MAB5014
10741-034EA 329 USD
10741-034
Anti-TFRC Mouse Monoclonal Antibody (Biotin) [clone: MEM-75]
Antibodies
Mouse monoclonal antibody raised against native TFRC.

Recommended Dilutions: Flow Cytometry: 1:200

Type: Primary
Antigen: TFRC
Clonality: Monoclonal
Clone: MEM-75
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR