Anti-CD22 Mouse Monoclonal Antibody (Biotin) [clone: MEM-01]

Supplier: Abnova
MAB5032
10706-242EA 329 USD
10706-242
Anti-CD22 Mouse Monoclonal Antibody (Biotin) [clone: MEM-01]
Antibodies
Mouse monoclonal antibody raised against native CD22.

Recommended Dilutions: Flow Cytometry: 1:500

Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone: MEM-01
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Primates
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR