Anti-MUC12 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12553
10726-216EA 638.29 USD
10726-216
Anti-MUC12 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of MUC12.

Recommended Dilutions: ELISA: 1:500-1:1000; Immunofluorescence: 1:500-1:1000; Immunohistochemistry (Frozen sections): 1:500-1:1000; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:500-1:1000; Immunoprecipitation: 1:500-1:1000; Western Blot: 1:500-1:1000

Type: Primary
Antigen: MUC12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR