Anti-ICAM3 Mouse Monoclonal Antibody (Biotin) [clone: MEM-171]

Supplier: Abnova
MAB5036
10721-192EA 329 USD
10721-192
Anti-ICAM3 Mouse Monoclonal Antibody (Biotin) [clone: MEM-171]
Antibodies
Mouse monoclonal antibody raised against native ICAM3.

Recommended Dilutions: Flow Cytometry: 1:500

Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: MEM-171
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR