Anti-LRP2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20330
10723-978EA 428.55 USD
10723-978
Anti-LRP2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant LRP2.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: LRP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR