Anti-GLCCI1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB20331
10715-932EA 428.55 USD
10715-932
Anti-GLCCI1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant GLCCI1.

Recommended Dilutions: Immunohistochemistry: 1:20-1:50

Type: Primary
Antigen: GLCCI1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR